Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1H,1H-Nonafluoropentylamine, TCI America™
SDP

Catalog No. N08721G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N08721G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N08721G Supplier TCI America Supplier No. N08721G

Chemical Identifiers

CAS 355-27-1
Molecular Formula C5H4F9N
Molecular Weight (g/mol) 249.08
MDL Number MFCD06213670
InChI Key SUNUNYBGDFIAME-UHFFFAOYSA-N
Synonym 1H,1H-Perfluoropentylamine
PubChem CID 2769669
IUPAC Name 2,2,3,3,4,4,5,5,5-nonafluoropentan-1-amine
SMILES NCC(F)(F)C(F)(F)C(F)(F)C(F)(F)F

Specifications

Color Yellow
UN Number 2735
Quantity 1 g
Formula Weight 249.08
Percent Purity ≥95.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1H,1H-Nonafluoropentylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.