accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 35502-06-8

IUPAC Name: 3-chloro-1,1-dimethoxypropane
Synonyms: 3-chloro-1,1-dimethoxypropane
Molecular Weight (g/mol): 138.59
Molecular Formula: C5H11ClO2
InChi Key: DXWRNRRBDAQWDB-UHFFFAOYSA-N
SMILES: COC(CCCl)OC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results