Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloropropionaldehyde Dimethyl Acetal 93.0+%, TCI America™
SDP

Catalog No. C24585G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C24585G 5 g
C245825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C24585G Supplier TCI America Supplier No. C24585G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 35502-06-8
Molecular Formula C5H11ClO2
Molecular Weight (g/mol) 138.591
MDL Number MFCD16038653
InChI Key DXWRNRRBDAQWDB-UHFFFAOYSA-N
Synonym 1-Chloro-3,3-dimethoxypropane
PubChem CID 142015
IUPAC Name 3-chloro-1,1-dimethoxypropane
SMILES COC(CCCl)OC

Specifications

Color Colorless
Boiling Point 45°C
Quantity 5 g
UN Number 1993
Formula Weight 138.59
Percent Purity ≥93.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Chloropropionaldehyde Dimethyl Acetal
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.