accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNH

CAS RN 35543-25-0

IUPAC Name: 1-(3-cyanopropyl)pyrrolidin-1-ium
Synonyms: 1-(3-cyanopropyl)pyrrolidin-1-ium
Molecular Weight (g/mol): 139.22
Molecular Formula: C8H15N2
InChi Key: OSDDDHPYSNZBPF-UHFFFAOYSA-O
SMILES: N#CCCC[NH+]1CCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results