accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OClOCH3

CAS RN 35573-93-4

IUPAC Name: 3-chloro-1,1-diethoxypropane
Synonyms: 3-chloro-1,1-diethoxypropane
Molecular Weight (g/mol): 166.65
Molecular Formula: C7H15ClO2
InChi Key: NXHONHDWVLPPCS-UHFFFAOYSA-N
SMILES: CCOC(CCCl)OCC
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (1)
  • (2)
  • (1)
  • (2)