accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSCH3

CAS RN 35599-83-8

IUPAC Name: 1-(tetradecylsulfanyl)tetradecane
Synonyms: 1-(tetradecylsulfanyl)tetradecane
Molecular Weight (g/mol): 426.83
Molecular Formula: C28H58S
InChi Key: OCTONCPZMJYQLP-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCSCCCCCCCCCCCCCC
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results