accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 35654-56-9

IUPAC Name: 4-chloro-6,7-dimethoxyquinoline
Synonyms: 4-chloro-6,7-dimethoxyquinoline
Molecular Weight (g/mol): 223.66
Molecular Formula: C11H10ClNO2
InChi Key: WRVHQEYBCDPZEU-UHFFFAOYSA-N
SMILES: COC1=C(OC)C=C2C(Cl)=CC=NC2=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results