Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-6,7-dimethoxyquinoline 98.0+%, TCI America™
SDP

Catalog No. C293825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C293825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C293825G Supplier TCI America Supplier No. C293825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 35654-56-9
Molecular Formula C11H10ClNO2
Molecular Weight (g/mol) 223.656
MDL Number MFCD07778437
InChI Key WRVHQEYBCDPZEU-UHFFFAOYSA-N
PubChem CID 459610
IUPAC Name 4-chloro-6,7-dimethoxyquinoline
SMILES COC1=CC2=C(C=CN=C2C=C1OC)Cl

Specifications

Melting Point 134°C
Color Yellow
Quantity 25 g
Formula Weight 223.66
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloro-6,7-dimethoxyquinoline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.