accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3566-44-7

IUPAC Name: hydrogen N1-(4-methoxyphenyl)benzene-1,4-diamine chloride
Synonyms: hydrogen N1-(4-methoxyphenyl)benzene-1,4-diamine chloride
Molecular Weight (g/mol): 250.73
Molecular Formula: C13H15ClN2O
InChi Key: HPQQXLXIGHOKNZ-UHFFFAOYSA-N
SMILES: [H+].[Cl-].COC1=CC=C(NC2=CC=C(N)C=C2)C=C1
Quantity 
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (2)
Color 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results