Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-4'-methoxydiphenylamine HCl, MP Biomedicals

Catalog No. ICN203561 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN203561 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN203561 Supplier MP Biomedicals, Inc Supplier No. 0520356101

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3566-44-7
Molecular Formula C13H15ClN2O
Molecular Weight (g/mol) 250.726
InChI Key HPQQXLXIGHOKNZ-UHFFFAOYSA-N
Synonym variamine blue b, unii-e365pn4l70, n-4-methoxyphenyl-1,4-benzenediamine hydrochloride, n-4-methoxyphenyl benzene-1,4-diamine monohydrochloride, 1,4-benzenediamine, n-4-methoxyphenyl-, monohydrochloride, n-4-methoxyphenyl-p-phenylenediamine hydrochloride, 1,4-benzenediamine, n1-4-methoxyphenyl-, hydrochloride 1:1, 4-amino-4'-methoxydiphenylamine hydrochloride, n-p-methoxyphenyl-p-phenylenediamine hydrochloride, 4-aminophenyl 4-methoxyphenyl amine, chloride
PubChem CID 77108
IUPAC Name 4-N-(4-methoxyphenyl)benzene-1,4-diamine;hydrochloride
SMILES COC1=CC=C(C=C1)NC2=CC=C(C=C2)N.Cl

Specifications

Quantity 1 g
Formula Weight 250.74
Chemical Name or Material 4-Amino-4«-Methoxydiphenylamine HCl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.