accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SOOOCl

CAS RN 3570-58-9

IUPAC Name: 2-chloroethyl methanesulfonate
Synonyms: 2-chloroethyl methanesulfonate
Molecular Weight (g/mol): 158.6
Molecular Formula: C3H7ClO3S
InChi Key: FMMYTRQXHORTCU-UHFFFAOYSA-N
SMILES: CS(=O)(=O)OCCCl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results