Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloroethyl Methanesulfonate 98.0+%, TCI America™
SDP

Catalog No. C22315G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C22315G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C22315G Supplier TCI America Supplier No. C22315G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3570-58-9
Molecular Formula C3H7ClO3S
Molecular Weight (g/mol) 158.60
MDL Number MFCD00007560
InChI Key FMMYTRQXHORTCU-UHFFFAOYSA-N
Synonym 2-chloroethylmethanesulfonate, methanesulfonic acid, 2-chloroethyl ester, chloroethyl methanesulfonate, chloroethyl methanesulphonate, 2-chloroethylmethanesulfonate, puriss, 2-chloroethyl methanesulphonate, beta-chloroethylmethanesulfonate, ethanol, 2-chloro-, methanesulfonate, methanesulfonic acid, chloroethyl ester, ethanol, 2-chloro-,1-methanesulfonate
PubChem CID 19128
IUPAC Name 2-chloroethyl methanesulfonate
SMILES CS(=O)(=O)OCCCl

Specifications

Melting Point 6°C
Color Colorless
Boiling Point 124°C
Quantity 5 g
Formula Weight 158.60
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Chloroethyl Methanesulfonate
RTECSNumber KK1960000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.