accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 35822-58-3

IUPAC Name: 1-bromo-2-(diethoxymethyl)benzene
Synonyms: 1-bromo-2-(diethoxymethyl)benzene
Molecular Weight (g/mol): 259.14
Molecular Formula: C11H15BrO2
InChi Key: APDGYQVYBWGBSD-UHFFFAOYSA-N
SMILES: CCOC(OCC)C1=CC=CC=C1Br
Boiling Point 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)