Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromobenzaldehyde Diethyl Acetal 97.0+%, TCI America™
SDP

Catalog No. B380025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B380025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B380025G Supplier TCI America Supplier No. B380025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 35822-58-3
Molecular Formula C11H15BrO2
Molecular Weight (g/mol) 259.14
MDL Number MFCD01075696
InChI Key APDGYQVYBWGBSD-UHFFFAOYSA-N
Synonym 2-bromobenzaldehyde diethyl acetal, 1-bromo-2-diethoxymethyl benzene, 2-diethoxymethyl bromobenzene, 2-bromobenzaldehydediethylacetal, benzene, 1-bromo-2-diethoxymethyl, 2-bromophenyl diethoxymethane, 2-bromo-alpha,alpha-diethoxytoluene, pubchem5370, acmc-1afuv, bromobenzaldehyde diethylacetal
PubChem CID 3482778
IUPAC Name 1-bromo-2-(diethoxymethyl)benzene
SMILES CCOC(OCC)C1=CC=CC=C1Br

Specifications

Color Yellow
Boiling Point 116°C
Quantity 25 g
Formula Weight 259.14
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromobenzaldehyde Diethyl Acetal
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.