Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromobenzaldehyde diethyl acetal, 98%, Thermo Scientific™

Catalog No. AAL1551306
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1551306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1551306 Supplier Thermo Scientific Chemicals Supplier No. L1551306

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35822-58-3
Molecular Formula C11H15BrO2
Molecular Weight (g/mol) 259.14
MDL Number MFCD01075696
InChI Key APDGYQVYBWGBSD-UHFFFAOYSA-N
Synonym 2-bromobenzaldehyde diethyl acetal, 1-bromo-2-diethoxymethyl benzene, 2-diethoxymethyl bromobenzene, 2-bromobenzaldehydediethylacetal, benzene, 1-bromo-2-diethoxymethyl, 2-bromophenyl diethoxymethane, 2-bromo-alpha,alpha-diethoxytoluene, pubchem5370, acmc-1afuv, bromobenzaldehyde diethylacetal
PubChem CID 3482778
IUPAC Name 1-bromo-2-(diethoxymethyl)benzene
SMILES CCOC(OCC)C1=CC=CC=C1Br

Specifications

Density 1.285
Boiling Point 95°C (1 mmHg)
Flash Point 113°C (235°F)
Refractive Index 1.515
Quantity 5 g
Beilstein 5935613
Formula Weight 259.14
Percent Purity 98%
Chemical Name or Material 2-Bromobenzaldehyde diethyl acetal
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.