accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONHNO

CAS RN 3589-41-1

IUPAC Name: benzyl N-(cyanomethyl)carbamate
Synonyms: benzyl N-(cyanomethyl)carbamate
Molecular Weight (g/mol): 190.2
Molecular Formula: C10H10N2O2
InChi Key: DVUXKEFDAGQPQU-UHFFFAOYSA-N
SMILES: O=C(NCC#N)OCC1=CC=CC=C1
Grade 
  • (2)
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)