accessibility menu, dialog, popup

UserName

Viewing profile as user:
N(R)N(R)O(S)(S)NNOHNH2HONHOH3C

CAS RN 35920-39-9

IUPAC Name: (2S,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-N-ethyl-3,4-dihydroxyoxolane-2-carboxamide
Synonyms: NECA
Molecular Weight (g/mol): 308.3
Molecular Formula: C12H16N6O4
InChi Key: JADDQZYHOWSFJD-AHTPIVMJNA-N
SMILES: CCNC(=O)[C@H]1O[C@H]([C@H](O)[C@@H]1O)N1C=NC2=C(N)N=CN=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)