accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 35951-28-1

IUPAC Name: 2-chlorocyclododecan-1-one
Synonyms: 2-chlorocyclododecan-1-one
Molecular Weight (g/mol): 216.75
Molecular Formula: C12H21ClO
InChi Key: VDNDILHUSVTMNI-UHFFFAOYNA-N
SMILES: ClC1CCCCCCCCCCC1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results