accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 36015-19-7

IUPAC Name: (2E)-3-(2-chloro-5-nitrophenyl)prop-2-enoate
Synonyms: (2E)-3-(2-chloro-5-nitrophenyl)prop-2-enoate
Molecular Weight (g/mol): 226.59
Molecular Formula: C9H5ClNO4
InChi Key: IHTLKLSRRSWIGD-DAFODLJHSA-M
SMILES: [O-]C(=O)\C=C\C1=CC(=CC=C1Cl)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results