CAS RN 36067-72-8
IUPAC Name:
dihydrogen 6-ethyl-4H,5H,6H,7H,8H-[1,3]oxazolo[4,5-d]azepin-2-amine dichloride
Synonyms:
dihydrogen 6-ethyl-4H,5H,7H,8H-[1,3]oxazolo[4,5-d]azepin-2-amine dichloride
Molecular Weight (g/mol):
254.16
Molecular Formula:
C9H17Cl2N3O
InChi Key:
HBLPYIOKPJVFQW-UHFFFAOYSA-N
SMILES:
[H+].[H+].[Cl-].[Cl-].CCN1CCC2=C(CC1)N=C(N)O2
1
–
1
of
1
results
1
R&D Systems™ B-HT 933 dihydrochloride
CAS: 36067-72-8 Molecular Formula: C9H17Cl2N3O