accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3607-17-8

IUPAC Name: (3-bromopropyl)triphenylphosphanium bromide
Synonyms: (3-bromopropyl)triphenylphosphanium bromide
Molecular Weight (g/mol): 464.18
Molecular Formula: C21H21Br2P
InChi Key: ZAHUZZUGJRPGKW-UHFFFAOYSA-M
SMILES: [Br-].BrCCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (3)
Percent Purity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)