Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(3-Bromopropyl)triphenylphosphonium bromide, 98%, Thermo Scientific™

Catalog No. AAA1304114
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1304114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1304114 Supplier Thermo Scientific Chemicals Supplier No. A1304114

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3607-17-8
Molecular Formula C21H21Br2P
Molecular Weight (g/mol) 464.181
MDL Number MFCD00011866
InChI Key ZAHUZZUGJRPGKW-UHFFFAOYSA-M
Synonym 3-bromopropyl triphenylphosphonium bromide, 3-bromopropyltriphenylphosphonium bromide, bromo 3-bromopropyl triphenylphosphorus, phosphonium, 3-bromopropyl triphenyl-, bromide, 3-bromopropyl triphenylphosphanium bromide, 3-bromopropyl triphenylphosphonium, acmc-209ikr, bromo 3-bromopropyl triphenylphosphorane, 3-brompropyl triphenylphosphonium bromide
PubChem CID 2723862
IUPAC Name 3-bromopropyl(triphenyl)phosphanium;bromide
SMILES C1=CC=C(C=C1)[P+](CCCBr)(C2=CC=CC=C2)C3=CC=CC=C3.[Br-]

Specifications

Melting Point 225°C to 229°C
Quantity 25 g
Beilstein 6422974
Sensitivity Hygroscopic
Formula Weight 464.19
Percent Purity 98%
Chemical Name or Material (3-Bromopropyl)triphenylphosphonium bromide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 222-770-9
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.