Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromopropyltriphenylphosphonium Bromide 98.0+%, TCI America™
SDP

Catalog No. B120825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B120825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B120825G Supplier TCI America Supplier No. B120825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3607-17-8
Molecular Formula C21H21Br2P
Molecular Weight (g/mol) 464.181
MDL Number MFCD00011866
InChI Key ZAHUZZUGJRPGKW-UHFFFAOYSA-M
Synonym 3-bromopropyl triphenylphosphonium bromide, 3-bromopropyltriphenylphosphonium bromide, bromo 3-bromopropyl triphenylphosphorus, phosphonium, 3-bromopropyl triphenyl-, bromide, 3-bromopropyl triphenylphosphanium bromide, 3-bromopropyl triphenylphosphonium, acmc-209ikr, bromo 3-bromopropyl triphenylphosphorane, 3-brompropyl triphenylphosphonium bromide
PubChem CID 2723862
IUPAC Name 3-bromopropyl(triphenyl)phosphanium;bromide
SMILES C1=CC=C(C=C1)[P+](CCCBr)(C2=CC=CC=C2)C3=CC=CC=C3.[Br-]

Specifications

Melting Point 226°C
Color White
Quantity 25 g
Formula Weight 464.18
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromopropyltriphenylphosphonium Bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.