accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOOCH3

CAS RN 36076-26-3

IUPAC Name: ethyl 2-[4-(2-ethoxy-2-oxoethyl)phenyl]acetate
Synonyms: ethyl 2-[4-(2-ethoxy-2-oxoethyl)phenyl]acetate
Molecular Weight (g/mol): 250.29
Molecular Formula: C14H18O4
InChi Key: JRSOAUOHTHOAOP-UHFFFAOYSA-N
SMILES: CCOC(=O)CC1=CC=C(CC(=O)OCC)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)