accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NHSOO

CAS RN 3622-84-2

IUPAC Name: N-butylbenzenesulfonamide
Synonyms: N-butyl-benzenesulfonamide plasthall bsa BM 4 (sulfonamide) dellatol bbs N-butylbenzene sulfonamide uniplex 214 plastomoll bmb cetamoll bmb
Molecular Weight (g/mol): 213.3
Molecular Formula: C10H15NO2S
InChi Key: IPRJXAGUEGOFGG-UHFFFAOYSA-N
SMILES: CCCCNS(=O)(=O)C1=CC=CC=C1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results