accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3637-31-8

IUPAC Name: 4,4,5,5,5-pentafluoropentanoic acid
Synonyms: 4,4,5,5,5-pentafluoropentanoic acid
Molecular Weight (g/mol): 192.09
Molecular Formula: C5H5F5O2
InChi Key: HHSIXPRDVJARSI-UHFFFAOYSA-N
SMILES: OC(=O)CCC(F)(F)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results