accessibility menu, dialog, popup

UserName

Viewing profile as user:
HNNNSOOONa(E/Z)

CAS RN 36402-77-4

IUPAC Name: sodium 4-{2-[4-(benzylamino)phenyl]diazen-1-yl}benzene-1-sulfonate
Synonyms: sodium 4-{2-[4-(benzylamino)phenyl]diazen-1-yl}benzenesulfonate
Molecular Weight (g/mol): 389.4
Molecular Formula: C19H16N3NaO3S
InChi Key: JGCBNZLRTUKUAQ-UHFFFAOYSA-M
SMILES: [Na+].[O-]S(=O)(=O)C1=CC=C(C=C1)N=NC1=CC=C(NCC2=CC=CC=C2)C=C1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results