accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 36410-81-8

IUPAC Name: (4-isothiocyanato-2,2,6,6-tetramethylpiperidin-1-yl)oxidanyl
Synonyms: 4-isothiocyanato-2,2,6,6-tetramethylpiperidin-1-yloxidanyl
Molecular Weight (g/mol): 213.32
Molecular Formula: C10H17N2OS
InChi Key: BKENZAFIBRCMKD-UHFFFAOYSA-N
SMILES: CC1(C)CC(CC(C)(C)N1[O])N=C=S
Quantity 
  • (1)
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)