Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Isothiocyanato-TEMPO, Free Radical 95%, Thermo Scientific™

Catalog No. AC290310250
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC290310250 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC290310250 Supplier Thermo Scientific Chemicals Supplier No. 290310250

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36410-81-8
Molecular Formula C10H17N2OS
Molecular Weight (g/mol) 213.32
MDL Number MFCD00010109
InChI Key BKENZAFIBRCMKD-UHFFFAOYSA-N
Synonym 4-isothiocyanato-2,2,6,6-tetramethylpiperidine 1-oxyl, 4-isothiocyanato-tempo, 4-isothiocyanato-2,2,6,6-tetramethylpiperidinooxyl, 4-itc-tempo, 4-isothiocyanato-1, 4-isothiocyanato-2,2,6,6-tetramethyl-1-piperidinyloxy, 4-isothiocyanato-2,2,6,6-tetramethylpiperidinooxy radical, 1-piperidinyloxy, 4-isothiocyanato-2,2,6,6-tetramethyl, 4-isothiocyanato-tempo, for esr-spectroscopy, 4-isothiocyanato-2,2,6,6-tetramethyl-1-piperidinyloxy, free radical
PubChem CID 147645
IUPAC Name (4-isothiocyanato-2,2,6,6-tetramethylpiperidin-1-yl)oxidanyl
SMILES CC1(C)CC(CC(C)(C)N1[O])N=C=S

Specifications

CAS Min % 94.0
CAS Max % 100.0
Quantity 25 mg
Formula Weight 213.32
Chemical Name or Material 4-Isothiocyanato-TEMPO, 95%
missing translation for 'healthHazard1' MOISTURE SENSITIVE
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.