accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3NSO

CAS RN 36415-21-1

IUPAC Name: 2-{[(furan-2-yl)methyl]sulfanyl}ethan-1-aminium
Synonyms: 2-[(furan-2-ylmethyl)sulfanyl]ethanaminium
Molecular Weight (g/mol): 158.24
Molecular Formula: C7H12NOS
InChi Key: VSPDYEHKAMKDNW-UHFFFAOYSA-O
SMILES: [NH3+]CCSCC1=CC=CO1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results