accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3648-36-0

ClH3CNClNH3CCH3H3C(E/Z)

CAS RN 3648-36-0

IUPAC Name: 2-(2-{4-[(2-chloroethyl)(methyl)amino]phenyl}ethenyl)-1,3,3-trimethyl-3H-indol-1-ium chloride
Synonyms: 2-(2-{4-[(2-chloroethyl)(methyl)amino]phenyl}ethenyl)-1,3,3-trimethylindol-1-ium chloride
Molecular Weight (g/mol): 389.36
Molecular Formula: C22H26Cl2N2
InChi Key: ZTBANYZVKCGOKD-UHFFFAOYSA-M
SMILES: [Cl-].CN(CCCl)C1=CC=C(C=CC2=[N+](C)C3=CC=CC=C3C2(C)C)C=C1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results