accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3PCH3SPCH3CH3S

CAS RN 3676-97-9

IUPAC Name: tetramethyl-1λ⁵,2λ⁵-diphosphane-1,2-dithione
Synonyms: tetramethyl-1λ⁵,2λ⁵-diphosphane-1,2-dithione
Molecular Weight (g/mol): 186.21
Molecular Formula: C4H12P2S2
InChi Key: LGZQQMXQPAHRIH-UHFFFAOYSA-N
SMILES: CP(C)(=S)P(C)(C)=S
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)