Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetramethyldiphosphine Disulfide, TCI America™
SDP

Catalog No. T13085G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T13085G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T13085G Supplier TCI America Supplier No. T13085G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3676-97-9
Molecular Formula C4H12P2S2
Molecular Weight (g/mol) 186.208
MDL Number MFCD00014459
InChI Key LGZQQMXQPAHRIH-UHFFFAOYSA-N
Synonym tetramethyldiphosphine disulfide, tetramethylbiphosphine disulfide, diphosphine, tetramethyl-, 1,2-disulfide, 1,1,2,2-tetramethyldiphosphane 1,2-disulfide, dimethylphosphinothioyl-dimethyl-thioxo-$l^ 5-phosphane, tetramethyl-1??,2??-diphosphane-1,2-dithione, diphosphine, tetramethyl-, disulfide, teramethyldiphosphine disulphide, tetramethyldiphosphoryl disulfide, 1,2-bisthiotetramethyldiphosphorane
PubChem CID 96660
IUPAC Name dimethylphosphinothioyl-dimethyl-sulfanylidene-$l^{5}-phosphane
SMILES CP(=S)(C)P(=S)(C)C

Specifications

Melting Point 226°C
Color White
Quantity 5 g
Formula Weight 186.21
Physical Form Crystalline Powder
Chemical Name or Material Tetramethyldiphosphine Disulfide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.