Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetramethyl Diphosphine Disulfide, MP Biomedicals

Catalog No. ICN222499 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN222499 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN222499 Supplier MP Biomedicals, Inc Supplier No. 0522249983
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3676-97-9
Molecular Formula C4H12P2S2
Molecular Weight (g/mol) 186.208
InChI Key LGZQQMXQPAHRIH-UHFFFAOYSA-N
Synonym tetramethyldiphosphine disulfide, tetramethylbiphosphine disulfide, diphosphine, tetramethyl-, 1,2-disulfide, 1,1,2,2-tetramethyldiphosphane 1,2-disulfide, dimethylphosphinothioyl-dimethyl-thioxo-$l^ 5-phosphane, tetramethyl-1??,2??-diphosphane-1,2-dithione, diphosphine, tetramethyl-, disulfide, teramethyldiphosphine disulphide, tetramethyldiphosphoryl disulfide, 1,2-bisthiotetramethyldiphosphorane
PubChem CID 96660
IUPAC Name dimethylphosphinothioyl-dimethyl-sulfanylidene-$l^{5}-phosphane
SMILES CP(=S)(C)P(=S)(C)C

Specifications

Quantity 250 mg
Formula Weight 185.21
Chemical Name or Material Tetramethyl Diphosphine Disulfide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.