accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 368-72-9

IUPAC Name: 3-amino-4-chlorobenzene-1-sulfonyl fluoride
Synonyms: 3-amino-4-chlorobenzenesulfonyl fluoride
Molecular Weight (g/mol): 209.62
Molecular Formula: C6H5ClFNO2S
InChi Key: HNZYBSQZUOJCDN-UHFFFAOYSA-N
SMILES: NC1=CC(=CC=C1Cl)S(F)(=O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results