Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-4-chlorobenzenesulfonyl Fluoride 99+%, Thermo Scientific™

Catalog No. AC162180050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC162180050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC162180050 Supplier Thermo Scientific Chemicals Supplier No. 162180050

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 368-72-9
Molecular Formula C6H5ClFNO2S
Molecular Weight (g/mol) 209.619
InChI Key HNZYBSQZUOJCDN-UHFFFAOYSA-N
Synonym 3-amino-4-chlorobenzenesulphonyl fluoride, benzenesulfonyl fluoride, 3-amino-4-chloro, 3-amino-4-chlorophenyl fluorosulfone, 3-amino-4-chlorophenylsulfonyl fluoride, benzenesulfonylfluoride, 3-amino-4-chloro, 3-amino-4-chloro benzene sulphonyl fluoride, 3-amino-4-chlorobenzene-1-sulfonyl fluoride, 3-amino-4-chlorobenzenesulfonic acid fluoride, 3-amino-4-chlorobenzene sulfonyl fluoride, 3-amino-4-chloro-benzenesulfonic acid fluoride
PubChem CID 97383
IUPAC Name 3-amino-4-chlorobenzenesulfonyl fluoride
SMILES C1=CC(=C(C=C1S(=O)(=O)F)N)Cl

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 64°C to 67°C
Color Brown
Infrared Spectrum Authentic
Linear Formula H2NC6H3(Cl)SO2F
Quantity 5 g
Formula Weight 209.62
Percent Purity ≥99.0%
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-4-chlorobenzenesulfonyl fluoride, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.