accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOOOOHOHHOOHHO

CAS RN 36804-17-8

IUPAC Name: 3,5,7-trihydroxy-2-[3-(4-hydroxy-3-methoxyphenyl)-2-(hydroxymethyl)-2,3-dihydro-1,4-benzodioxin-6-yl]-3,4-dihydro-2H-1-benzopyran-4-one
Synonyms: silymarin silybine hevert brand of silybin hepar-pasc silliver merck dura brand of silybin pascoe brand of silybin silybum marianum fruit extract silibininum silibin
Molecular Weight (g/mol): 482.44
Molecular Formula: C25H22O10
InChi Key: SEBFKMXJBCUCAI-UHFFFAOYNA-N
SMILES: COC1=C(O)C=CC(=C1)C1OC2=C(OC1CO)C=CC(=C2)C1OC2=CC(O)=CC(O)=C2C(=O)C1O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results