accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3699-01-2

IUPAC Name: 1-methyl-4-(phenylsulfanyl)benzene
Synonyms: 1-methyl-4-(phenylsulfanyl)benzene
Molecular Weight (g/mol): 200.3
Molecular Formula: C13H12S
InChi Key: CPZFPNKPHBCUOB-UHFFFAOYSA-N
SMILES: CC1=CC=C(SC2=CC=CC=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results