accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaCH3SiH3CCH3OO

CAS RN 37013-20-0

IUPAC Name: sodium 3-(trimethylsilyl)propanoate
Synonyms: sodium 3-(trimethylsilyl)propanoate
Molecular Weight (g/mol): 168.24
Molecular Formula: C6H13NaO2Si
InChi Key: OIIWPAYIXDCDNL-UHFFFAOYSA-M
SMILES: [Na+].C[Si](C)(C)CCC([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results