accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOHNO

CAS RN 37033-95-7

IUPAC Name: ethyl 5-(benzyloxy)-1H-indole-2-carboxylate
Synonyms: ethyl 5-(benzyloxy)-1H-indole-2-carboxylate
Molecular Weight (g/mol): 295.34
Molecular Formula: C18H17NO3
InChi Key: DCIFXYFKVKDOLL-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC2=CC(OCC3=CC=CC=C3)=CC=C2N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results