accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONN(E/Z)

CAS RN 37075-25-5

IUPAC Name: 4-({[4-(pentyloxy)phenyl]methylidene}amino)benzonitrile
Synonyms: 4-({[4-(pentyloxy)phenyl]methylidene}amino)benzonitrile
Molecular Weight (g/mol): 292.38
Molecular Formula: C19H20N2O
InChi Key: IUWUDHSOMQRZCV-UHFFFAOYSA-N
SMILES: CCCCCOC1=CC=C(C=NC2=CC=C(C=C2)C#N)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results