accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOF

CAS RN 371-49-3

IUPAC Name: butyl 2-fluoroacetate
Synonyms: butyl 2-fluoroacetate
Molecular Weight (g/mol): 134.15
Molecular Formula: C6H11FO2
InChi Key: FZXXGBGMLALCIH-UHFFFAOYSA-N
SMILES: CCCCOC(=O)CF

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results