accessibility menu, dialog, popup

UserName

Viewing profile as user:
HO

CAS RN 37275-49-3

IUPAC Name: tricyclo[5.2.1.0²,⁶]dec-4-en-8-ol
Synonyms: tricyclo[5.2.1.0²,⁶]dec-4-en-8-ol
Molecular Weight (g/mol): 150.22
Molecular Formula: C10H14O
InChi Key: LDUKQFUHJZHLRC-UHFFFAOYNA-N
SMILES: OC1CC2CC1C1C=CCC21
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results