accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3737-41-5

IUPAC Name: 3,3,4,4-tetrachloro-1λ⁶-thiolane-1,1-dione
Synonyms: 3,3,4,4-tetrachloro-1λ⁶-thiolane-1,1-dione
Molecular Weight (g/mol): 257.93
Molecular Formula: C4H4Cl4O2S
InChi Key: GCAXGCSCRRVVLF-UHFFFAOYSA-N
SMILES: ClC1(Cl)CS(=O)(=O)CC1(Cl)Cl
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results