accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3739-64-8

IUPAC Name: 1-(prop-2-en-1-yloxy)butane
Synonyms: 1-(prop-2-en-1-yloxy)butane
Molecular Weight (g/mol): 114.19
Molecular Formula: C7H14O
InChi Key: IBTLFDCPAJLATQ-UHFFFAOYSA-N
SMILES: CCCCOCC=C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results