Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Allyl Butyl Ether 97.0+%, TCI America™
SDP

Catalog No. A148125ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A148125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A148125ML Supplier TCI America Supplier No. A148125ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3739-64-8
Molecular Formula C7H14O
Molecular Weight (g/mol) 114.19
MDL Number MFCD00009460
InChI Key IBTLFDCPAJLATQ-UHFFFAOYSA-N
Synonym allyl butyl ether, ether, allyl butyl, butyl allyl ether, allyl n-butyl ether, 1-allyloxy butane, butane, 1-2-propenyloxy, allyln-butylether, acmc-1cncm, 3-butoxy-1-propene #
PubChem CID 77332
IUPAC Name 1-(prop-2-en-1-yloxy)butane
SMILES CCCCOCC=C

Specifications

Color Yellow
Boiling Point 120°C
Quantity 25 mL
UN Number 3271
Formula Weight 114.19
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Allyl Butyl Ether
EINECSNumber (2)-0373&(9)-0128
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.