accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3742-34-5

IUPAC Name: ethenylcyclopentane
Synonyms: ethenylcyclopentane
Molecular Weight (g/mol): 96.17
Molecular Formula: C7H12
InChi Key: BEFDCLMNVWHSGT-UHFFFAOYSA-N
SMILES: C=CC1CCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results