accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COONH2H2N

CAS RN 37466-90-3

IUPAC Name: ethyl 3,4-diaminobenzoate
Synonyms: ethyl 3,4-diaminobenzoate
Molecular Weight (g/mol): 180.21
Molecular Formula: C9H12N2O2
InChi Key: NUJBTXFFJUGENN-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC=C(N)C(N)=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results