accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 376348-65-1

IUPAC Name: 4,4-difluoro-N-[(1S)-3-[(1R,5S)-3-[3-methyl-5-(propan-2-yl)-4H-1,2,4-triazol-4-yl]-8-azabicyclo[3.2.1]octan-8-yl]-1-phenylpropyl]cyclohexane-1-carboxamide
Synonyms: selzentry MVC celsentri
Molecular Weight (g/mol): 513.68
Molecular Formula: C29H41F2N5O
InChi Key: GSNHKUDZZFZSJB-HLMSNRGBSA-N
SMILES: CC(C)C1=NN=C(C)N1C1C[C@@H]2CC[C@H](C1)N2CC[C@H](NC(=O)C1CCC(F)(F)CC1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)