accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 376591-03-6

IUPAC Name: 5-(4-cyanophenyl)-2,3-bis(4-methylphenyl)-3H-1,2λ⁵,3,4-tetrazol-2-ylium chloride
Synonyms: 5-(4-cyanophenyl)-2,3-bis(4-methylphenyl)-1,2λ⁵,3,4-tetrazol-2-ylium chloride
Molecular Weight (g/mol): 387.87
Molecular Formula: C22H18ClN5
InChi Key: MEDRXJSBVPMSHT-UHFFFAOYSA-M
SMILES: [Cl-].CC1=CC=C(C=C1)N1N=C(N=[N+]1C1=CC=C(C)C=C1)C1=CC=C(C=C1)C#N
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results